missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MembranePro™ Functional Protein Support Kit

Product Code. p-7076104
Click to view available options
Sufficient For:
10 Reactions
Unit Size:
10 reactions
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 17183029

Brand: Invitrogen™ A11668

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

For customers studying GPCRs and other membrane proteins, MembranePro™ is the ready-to-use system that delivers enriched, functional membrane proteins efficiently and reliably.

Benefits of MembranePro™ include:

  • Proteins are displayed on human (or other mammalian) cell membranes
  • Cellular quality control and mammalian posttranslational processing helps produce functional protein
  • Proteins bud off as lipoparticles and are easily collected from the culture medium
  • Lipoparticles are enriched with expressed receptors

MembranePro™ workflow is less labor intensive in comparison to traditional production of membrane fractions. Briefly, the MembranePro™ protocol allows you to:

  1. Clone your gene of interest using a TOPO™ vector
  2. Transfect with Lipofectamine™ 2000 and MembranePro™ Reagent into 293FT cells
  3. Harvest lipoparticles 48h post-transfection, concentrate with MembranePro™ Precipitation Mix
  4. Store lipoparticles at -80 C or proceed to downstream assays

MembranePro™ products are offered in two configurations – as an expression kit (TOPO™ vector, Lipofectamine™ 2000, MembranePro™ Precipitation Mix, MembranePro™ Reagent, 293FT cells) and as a support kit (Lipofectamine™ 2000, MembranePro™ Precipitation Mix, MembranePro™ Reagent). The expression kit is only available in a 10 reaction size; the support kit is available in 10, 60 and 600 reaction sizes.

TRUSTED_SUSTAINABILITY

Specifications

Type MembranePro Functional Protein Support Kit
Sufficient For 10 Reactions
Delivery Type Virus-Like Particles (VLPs)
Quantity 10 reactions kit
Content And Storage •1 x 75 ml MembranePro™ Precipitation Mix (Store at room temperature)
•1 x 0.3 mg (1 mg⁄ml) MembranePro™ Reagent (Store at -20°C)
•3 x 0.75 ml Lipofectamine™ 2000 (Store at +4°C)
•Manual
Selection Agent (Eukaryotic) None
Expression Mechanism Cell-Based Expression
High-throughput Compatibility High-throughput Compatible
Expression System Mammalian
Expressed Protein Type Membrane Protein
Product Line MembranePro
For Use With (Application) Membrane Protein Expression
Promoter EF-1α
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.