Learn More
Fisherbrand™ Ion Selective Membrane Replacement Kits
Product Code. 11719728 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
83919 Device
FOR 83925
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11719728 83919 Device Each
11769728 FOR 83925 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11719728
  • ONLINE EXCLUSIVE PRICE
    358.74 EUR
    188.80 EUR / Pack of 1
    Save 169.94 EUR
    47% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
Sample preparation steps such as filtering and distillation are rarely necessary with this kit for the combined ion-sensitive electrode with a solid membrane.
  • Membrane replacement kit for ion-sensitive electrodes
  • For  83919 device
  • Allows for analysis time generally less than one minute
Specifications
Description Membrane replacement kit for ion-sensitive electrodes.
For Use With (Equipment) 83919 Device