Beschreibung
Gibco™ MEM Amino Acids Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Amino Acids Solution is formulated to contain 50X the essential amino acids (except L-glutamine) found in the standard Minimum Essential Medium (MEM). The complete formulation is available.
Product Use
For in vitro diagnostic use.
Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Amino Acids Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM Amino Acids Solution product made in our Grand Island facility (11130-051). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Product Use
For in vitro diagnostic use.
Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Amino Acids Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM Amino Acids Solution product made in our Grand Island facility (11130-051). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Spezifikation
Spezifikation
| Certifications/Compliance | ISO 13485 standard, FDA |
| Content And Storage | 2 to 8°C |
| Description | MEM Amino Acids Solution, 20 x 100mL, Liquid, 50X Concentration, Without Glutamine, Store at 2 to 8°C, Ambient Shipping Condition, 12 Months Shelf Life, For Mammalian Cell Culture |
| Concentration | 50 X |
| Form | Liquid |
| Product Type | Amino Acid Supplement |
| Without Additives | No Glutamine |
| Formulation | L-Arginine hydrochloride, L-cystine, L-histidine hydrochloride-H2O, L-isoleucine, L-leucine, L-lysine hydrochloride, L-methionine, L-phenylalanine, L-threonine, L-tryptophan, L-tyrosine, L-valine |
| For Use With (Application) | Mammalian Cell Culture |
| Quantity | 20 x 100 mL |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?