Learn More
Gibco™ MEM (10X), no glutamine
GREENER_CHOICE
Product Code. 11580566 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
5000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11580566 10 x 500 mL 5000mL
11570566 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11580566 Supplier Gibco™ Supplier No. 21430079
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
MEM (10X), no glutamine
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle’s salts.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Concentrated forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10 X
Form Liquid
Product Type MEM Medium
With Additives Phenol Red
Without Additives No Glutamine, No HEPES, No Sodium Bicarbonate
For Use With (Application) Mammalian cell culture
Quantity 10 x 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 1.2 to 1.8, 4.8 to 5.3
Regulatory Status In-vitro Diagnostic Use
Content And Storage 2 to 8°C
Description MEM Medium, 500mL, 10X Concentration, Liquid Form, With Phenol Red, 1.2 to 1.8, 4.8 to 5.3 pH, 12 Months Shelf Life, 2400 to 3000mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture