Learn More
Meleagrin, TRC
Product Code. 30138192
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138192 10 mg 10mg
30137311 5 mg 5mg
17997334 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138192 Supplier Toronto Research Chemicals Supplier No. TRCM21533510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71751-77-4
Molecular Formula C23 H23 N5 O4
Molecular Weight (g/mol) 433.46
Synonym (3E,7aR,12aS)-7a-(1,1-Dimethyl-2-propenyl)-7a,12-dihydro-6-hydroxy-3-(1H-imidazol-4-ylmethylene)-12-methoxy-1H,5H-imidazo[1',2':1,2]pyrido[2,3-b]indole-2,5(3H)-dione, [7aR-(3E,7aR*,12aS*)]-7a-(1,1-Dimethyl-2-propenyl)-7a,12-dihydro-6-hydroxy-3-(1H-imidazol-4-ylmethylene)-12-methoxy-1H,5H-imidazo[1',2':1,2]pyrido[2,3-b]indole-2,5(3H)-dione, 6-O-Demethyloxaline;
SMILES CON1c2ccccc2[C@@]3(C=C(O)C(=O)N4\C(=C\c5cnc[nH]5)\C(=O)N[C@@]134)C(C)(C)C=C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C23H23N5O4/c1-5-21(2,3)22-11-18(29)20(31)27-17(10-14-12-24-13-25-14)19(30)26-23(22,27)28(32-4)16-9-7-6-8-15(16)22/h5-13,29H,1H2,2-4H3,(H,24,25)(H,26,30)/b17-10+/t22-,23-/m0/s1
Formula Weight 433.175
Chemical Name or Material Meleagrin