missing translation for 'onlineSavingsMsg'
Learn More

Melatonin, MP Biomedicals

Product Code. 11423480 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423480 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423480 Supplier MP Biomedicals Supplier No. 02102254.1
  • ONLINE EXCLUSIVE PRICE
    43.00 EUR
    40.95 EUR / 100mg
    Save 2.05 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Melatonin is a hormone of the pineal gland

Specifications

CAS 73-31-4
Melting Point 114°C to 124°C
Color White
Molecular Formula C13H16N2O2
Linear Formula C13H16N2O2
MDL Number MFCD00005655
Quantity 100 mg
Synonym melatonin, melatonine, n-acetyl-5-methoxytryptamine, circadin, 5-methoxy-n-acetyltryptamine, n-2-5-methoxy-1h-indol-3-yl ethyl acetamide, melatol, melatonex, melovine
Solubility Information Soluble in methanol, ethanol (50mg/mL - clear to slightly hazy, colorless to faint yellow solution); Very slightly soluble in 0.1 N HCl (0.8mg/mL), 0.1 N NaOH (0.2mg/mL) or water (0.1mg/mL).
InChI Key DRLFMBDRBRZALE-UHFFFAOYSA-N
SMILES CC(=O)NCCC1=CNC2=C1C=C(C=C2)OC
IUPAC Name N-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide
Molecular Weight (g/mol) 232.283
PubChem CID 896
ChEBI CHEBI:16796
Formula Weight 232.28
Physical Form Powder
Chemical Name or Material MELATONIN
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.