missing translation for 'onlineSavingsMsg'
Learn More

Melamine Phosphate, TRC

Product Code. 30010985
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010985 25 g 25g
30009625 5 g 5g
30008220 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010985 Supplier Toronto Research Chemicals Supplier No. TRCM20872325G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20208-95-1
Molecular Formula C3 H6 N6 . H3 O4 P
Molecular Weight (g/mol) 224.115
Synonym 1,3,5-Triazine-2,4,6-triamine, phosphate (1:1), Melamine, phosphate (1:1) (8CI), 1,3,5-Triazinane-2,4,6-triimine phosphate, Amgard NH, M 850103, Melagard MP, Melamine monophosphate, Melamine phosphate
IUPAC Name phosphoric acid;1,3,5-triazine-2,4,6-triamine
SMILES Nc1nc(N)nc(N)n1.OP(=O)(O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C3H6N6.H3O4P/c4-1-7-2(5)9-3(6)8-1;1-5(2,3)4/h(H6,4,5,6,7,8,9);(H3,1,2,3,4)
Formula Weight 224.0423
Chemical Name or Material Melamine Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.