Learn More
STEMCELL Technologies MegaCult™-C Avidin-Alkaline Phosphatase Conjugate
Product Code. 17128241
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17128241 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17128241 Supplier STEMCELL Technologies Supplier No. 04905
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Avidin alkaline phosphatase conjugate
  • MegaCult™-C Staining Kit for CFU-Mk contains all the reagents needed to stain fixed and dehydrated CFU-Mk colonies cultured in MegaCult™-C collagen-based medium
  • Megakaryocytes and platelets expressing the glycoprotein IIb/IIIa are stained with anti-human CD41 antibody and an alkaline phosphatase detection system
  • The kit is also suitable for detection of bipotential erythroid-megakaryocyte progenitor cells (BFU-E/Mk)
Specifications
Quantity 200 μL
Product Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Colony Assay, Immunocytochemistry