missing translation for 'onlineSavingsMsg'
Learn More

Meclofenamic Acid, TRC

Product Code. 30136734
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136734 25 mg 25mg
30133286 10 mg 10mg
30011674 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136734 Supplier Toronto Research Chemicals Supplier No. TRCM20275025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644-62-2
Molecular Formula C14 H11 Cl2 N O2
Molecular Weight (g/mol) 296.15
Synonym Anthranilic acid, N-(2,6-dichloro-m-tolyl)- (7CI,8CI), 2-[(2,6-Dichloro-3-methylphenyl)amino]benzoic acid, Arquel, INF 4668, Meclofenamic acid, Meclophenamic acid, N-(2,6-Dichloro-3-methylphenyl)anthranilic acid, N-(2,6-Dichloro-m-tolyl)anthranilic acid, N-(3-Methyl-2,6-dichlorophenyl)anthranilic acid, NSC 95309, Meclofenamic Acid
IUPAC Name 2-(2,6-dichloro-3-methylanilino)benzoic acid
SMILES Cc1ccc(Cl)c(Nc2ccccc2C(=O)O)c1Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H11Cl2NO2/c1-8-6-7-10(15)13(12(8)16)17-11-5-3-2-4-9(11)14(18)19/h2-7,17H,1H3,(H,18,19)
Formula Weight 295.0167
Percent Purity >95%
Chemical Name or Material Meclofenamic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.