missing translation for 'onlineSavingsMsg'
Learn More

Meclofenamic Acid, Mikromol™

Product Code. 18082850
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18082850 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18082850 Supplier Mikromol Supplier No. MM3124.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Meclofenamic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Meclofenamic Acid
CAS 644-62-2
Analyte or Component Names Meclofenamic Acid
Shipping Condition Room Temperature
Molecular Formula C14 H11 Cl2 N O2
Certifications/Compliance ISO 17034
Synonym Benzoic acid, 2-[(2,6-dichloro-3-methylphenyl)amino]-, Anthranilic acid, N-(2,6-dichloro-m-tolyl)- (7CI,8CI), Arquel, INF 4668, Meclofenamic acid, Meclophenamic acid, N-(2,6-Dichloro-3-methylphenyl)anthranilic acid, N-(2,6-Dichloro-m-tolyl)anthranilic acid, N-(3-Methyl-2,6-dichlorophenyl)anthranilic acid, NSC 95309
InChI Formula InChI=1S/C14H11Cl2NO2/c1-8-6-7-10(15)13(12(8)16)17-11-5-3-2-4-9(11)14(18)19/h2-7,17H,1H3,(H,18,19)
SMILES Cc1ccc(Cl)c(Nc2ccccc2C(=O)O)c1Cl
IUPAC Name 2-(2,6-dichloro-3-methylanilino)benzoic acid
Molecular Weight (g/mol) 296.15
Quantity 50 mg
Formula Weight 295.0167 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.