Learn More
Meclofenamate Sodium, TRC
Product Code. 17972185
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972185 250 mg 250mg
17904015 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17972185 Supplier Toronto Research Chemicals Supplier No. TRCM202740250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6385-02-0
Molecular Formula C14 H10 Cl2 N O2 . Na
Molecular Weight (g/mol) 318.13
Synonym Benzoic acid, 2-[(2,6-dichloro-3-methylphenyl)amino]-, sodium salt (1:1), Anthranilic acid, N-(2,6-dichloro-m-tolyl)-, monosodium salt (8CI), Benzoic acid, 2-[(2,6-dichloro-3-methylphenyl)amino]-, monosodium salt (9CI), CI 583, Meclofenamate Sodium, Meclomen, N-(2,6-Dichloro-m-tolyl)anthranilic acid sodium salt, Sodium N-(2,6-dichloro-m-tolyl)anthranilate, Sodium meclofenamate, Sodium meclophenamate
IUPAC Name sodium;2-(2,6-dichloro-3-methylanilino)benzoate
SMILES [Na+].Cc1ccc(Cl)c(Nc2ccccc2C(=O)[O-])c1Cl
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C14H11Cl2NO2.Na/c1-8-6-7-10(15)13(12(8)16)17-11-5-3-2-4-9(11)14(18)19;/h2-7,17H,1H3,(H,18,19);/q;+1/p-1
Formula Weight 316.9986
Chemical Name or Material Meclofenamate Sodium