Learn More
TMC Hallcrest™ Thermax Chillchecker Temperature Indicators
Product Code. 11717218
Change view
Click to view available options
Display:
-17°C
-2°C
-8°C
9°C
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display unitSize
11717218 -8°C Pack of 100
11737218 -2°C Pack of 100
11767218 9°C Pack of 100
11707218 -17°C Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11717218 Supplier TMC Hallcrest™ Supplier No. M01KTHEN08C01X1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provide an irreversible identification for temperature controled issues. TMC Hallcrest™ Thermax Chillchecker Temperature Indicators can be used for transporation and storage of goods applications.
  • Confirmation that the temperatures have not been exceeded.
  • Color changes to blue when temperature is reached.
  • Easy to use and read.
  • Self Adhesive.
  • Available temperatures between -17 to 20°C.
  • Accurate to +/-1°C
Specifications
Depth (English) 0.23 in.
Depth (Metric) 6 mm
Disposable Single-use
Material PVC
Width (English) 1.25 in.
Product Type Temperature Indicator
Accuracy ±1°C
Display -8°C
Width (Metric) 32 mm