Learn More
MDBN, TRC
Specifications
Specifications
| Chemical Name or Material | MDBN |
| CAS | 1485-00-3 |
| Molecular Formula | C10 H13 N3 O2 |
| Quantity | 25 mg |
| Synonym | 5-(2-Nitroethenyl)-1,3-benzodioxole, 3,4-(Methylenedioxy)-β-nitro-styrene, 1-(2-Nitrovinyl)-3,4-methylenedioxybenzene, 3,4-(Methylenedioxy)-β-nitrostyrene, 3,4-Methylenedioxy-1-(2-nitrovinyl)benzene, 5-(2-Nitrovinyl)-1,3-benzodioxole, 5-(2-Nitrovinyl)benzodioxole, NSC 10120, NSC 105303, NSC 170724 |
| InChI Formula | InChI=1S/C10H13N3O2/c1-8(12(2)3)11-9-4-6-10(7-5-9)13(14)15/h4-7H,1-3H3/b11-8+ |
| SMILES | CN(C)\C(=N\c1ccc(cc1)[N+](=O)[O-])\C |
| IUPAC Name | N,N-dimethyl-N'-(4-nitrophenyl)ethanimidamide |
| Molecular Weight (g/mol) | 207.229 |
| Formula Weight | 207.101 |