Learn More
MDBN, TRC
Product Code. 30481578
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481578 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30481578 Supplier Toronto Research Chemicals Supplier No. TRCM19975050MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material MDBN
CAS 1485-00-3
Molecular Formula C10 H13 N3 O2
Quantity 50 mg
Synonym 5-(2-Nitroethenyl)-1,3-benzodioxole, 3,4-(Methylenedioxy)-β-nitro-styrene, 1-(2-Nitrovinyl)-3,4-methylenedioxybenzene, 3,4-(Methylenedioxy)-β-nitrostyrene, 3,4-Methylenedioxy-1-(2-nitrovinyl)benzene, 5-(2-Nitrovinyl)-1,3-benzodioxole, 5-(2-Nitrovinyl)benzodioxole, NSC 10120, NSC 105303, NSC 170724
InChI Formula InChI=1S/C10H13N3O2/c1-8(12(2)3)11-9-4-6-10(7-5-9)13(14)15/h4-7H,1-3H3/b11-8+
SMILES CN(C)\C(=N\c1ccc(cc1)[N+](=O)[O-])\C
IUPAC Name N,N-dimethyl-N'-(4-nitrophenyl)ethanimidamide
Molecular Weight (g/mol) 207.229
Formula Weight 207.101