Learn More
Novus Biologicals™ MCPIP1/ZC3H12A Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Specifications
Specifications
| Concentration | 0.2 mg/ml |
| For Use With (Application) | Antibody Competition |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| Gene ID (Entrez) | 80149 |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | Reg1, Regnase-1, zinc finger CCCH-type containing 12A |
| Gene Symbol | ZC3H12A |
| Research Category | Cancer, Cell Biology, Editing and Processing Endonucleases, Immunology, Inflammation |
| Protein | MCPIP1/ZC3H12A |