Learn More
Maxacalcitol-d6, MedChemExpress
Product Code. 16449047
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449047 5 mg 5mg
16411509 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16449047 Supplier Medchem Express Supplier No. HY15329
 Restricted Item
Estimated Shipment: 21-09-2026
MedChemExpress Maxacalcitol-D6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), which is a non-calcemic vitamin D3 analog and VDR ligand of VDR-like receptors.
Maxacalcitol-D6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), which is a non-calcemic vitamin D3 analog and VDR ligand of VDR-like receptors.
Specifications
Chemical Name or Material Maxacalcitol-d6
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C26H36D6O4
Color White
SMILES O[C@@H](C[C@H](O)C/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](OCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2
Molecular Weight (g/mol) 424.65
Quantity 5 mg
Formula Weight 424.65
Percent Purity 96.8%
Grade Research