Learn More
IKA™ MATRIX Orbital Delta FP
GREENER_CHOICE
Product Code. 16248681 Shop All IKA Products
Click to view available options
Orbit:
3 mm
Unit Size:
Each
Product Code. 16248681
Brand: IKA™ 0030000751
IKA MATRIX Orbital Delta FP mixes, heats and has a universal holder for microtiter plates. Whether its blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays – samples with even the smallest volumes mix reliably and completely in all laboratory applications.
  • Large and clear display with a convenient menu structure allows for intuitive operation
  • Stable quick lock for fast and easy change of inserts and attachments as well as automatic attachment detection
  • Speeds of up to 2,000 rpm and shaker diameter of 3 mm ensure the liquid mixes quickly and effectively
  • Two different speed modes for gentle heating of the samples
  • Programmable ramps allow for automatic execution of mixing and heating tasks
  • Smart speed adjustment according to insert, attachment and filling volume
  • Aluminum die cast housing ensures secure positioning, excellent stability and everyday robustness
  • No cross-contamination and an optimal mixing result.
TRUSTED_SUSTAINABILITY
Specifications
Orbit 3 mm
Load Bearing Capacity (Metric) 0.3 kg
Speed Control Accuracy 13 rpm Steps
Speed Range 0 to 2000 rpm
Temperature Range (Metric) Ambient +4° to 100°C
Plug Type EU Plug
Temperature Accuracy 0.5°C
Temperature Uniformity 0.5 (at 20°C to 45°C); 1.5°C
Frequency 50/60 Hz
Material Aluminum die-cast housing
Weight (Metric) 9.2 kg
Display TFT
Humidity Range 80%
Includes Power cable; USB cable
Platform Style Fixed attachment
Timer Range 0 to 6000 min.