Learn More
Fisherbrand™ Floor Mats
Product Code. 12816445 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
100 x 100 mm
170 x 160 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12816445 170 x 160 mm Each
12806445 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12816445
  • ONLINE EXCLUSIVE PRICE
    4.60 EUR
    3.59 EUR / Pack of 1
    Save 1.01 EUR
    21% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
Helps prevent fatigue when standing for long durations.
  • Polymeric compound
  • Available in different sizes
  • Blue in color
Specifications
Product Type Mat
Color Blue
Dimensions (L x W) 170 x 160 mm
Quantity 1 Ea.