Learn More
Masterflex™ L/S™ Compact Variable-Speed Pump Drive
Product Code. 11791349 Shop All Masterflex Products
Change view
Click to view available options
Electrical Requirements:
115/230 VAC 120 mA
12 VDC
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Unit Size
11791349 115/230 VAC 120 mA Each
11765429 12 VDC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11791349 Supplier Masterflex™ Supplier No. 7720020
  • ONLINE EXCLUSIVE PRICE
    1225.00 EUR
    1098.12 EUR / Pack of 1
    Save 126.88 EUR
    10% Off
Estimated Shipment: 07-12-2026
Use these drives when you need to pump fluid at a higher rate, but space is limited in your lab. MasterFlex™ L/S™ Compact Variable-Speed Pump Drive delivers versatile performance in limited space.
Drive accepts two different pump head types: Standard and Easy-Load™. Separate single-turn speed control and on/off switch with green power indicator maintain speed setting while turning pump on/off. Stackable, compact drive is perfect for small work areas.
  • Painted steel housing is IP22 rated. Housing protects drive from vertically falling water
  • MAX button runs pump at maximum rpm—use to prime or to rapidly flush tubing
  • Reversible permanent magnet DC motor lets you pump in either direction
  • Remote control options on the back of pump
  • Start/stop pump via contact closure
Specifications
Electrical Requirements 115/230 VAC 120 mA
Flow Rate 2.1 to 560 mL/min.
Overall Height (Metric) 114 mm
Overall Length (Metric) 13.6 cm
Overall Width (Metric) 132 mm
Reversible Yes
Speed Control ±5%
Type Pump Drive
Voltage 115/230 VAC
Speed 35–200 rpm