Learn More
Masterflex™ Chem-Durance™ Bio L/S™ High-Performance Precision Pump Tubing
Product Code. 11785648
Change view
Click to view available options
Tubing Size:
15
24
35
36
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size unitSize
11785648 36 Each
11816953 15 Each
11826953 24 Each
11836953 35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11785648 Supplier Masterflex™ Supplier No. 0644236
Add to basket
Add to basket
Use this tubing for a wide range of laboratory, pharmaceutical processing and biotech applications. Masterflex™ Chem-Durance™ Bio L/S™ High Performance Precision Pump Tubing features excellent chemical resistance and durability under pressure.
  • For applications involving pressure, suction lift, viscous fluids or long tubing life
  • Thicker wall compared to precision pump tubing
  • Performs in a wide range of chemical applications limiting the number of tubing formulations you need to keep on hand
  • Low spallation maintains purity of fluid
  • Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers and extending the life of the tubing
  • Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids Complies with USP Class VI, FDA, REACH, RoHS
  • Excellent chemical compatibility and biocompatibility
  • Excellent pump life and durability under pressure
  • Excellent chemical resistance and long pumping life
  • Meets USP Class VI requirements
Specifications
Autoclavable Autoclavable
Diameter (English) Inner 38/100 in.
Diameter (Metric) Inner 9.7 mm
Material Chem-Durance Bio
Overall Length (English) 50 ft.
Overall Length (Metric) 1520 cm
For Use With (Application) Laboratory, pharmaceutical processing and biotech applications
Min. Operating Temperature (English) -71°F
Min. Operating Temperature (Metric) -60°C
Tubing Size 36
Flow Rate 3400 mL/min.
Height (Metric) 15.2 mm Water
Height (English) 27 ft. Water
Color Opaque
Maximum Pressure 20 psi (1.4 bar)
Vacuum Level 610 mm Hg
Max. Operating Temperature (English) 165°F
Max. Operating Temperature (Metric) 74°C