Learn More
Eppendorf™ Mastercycler™ nexus eco, 230 V/50 – 60 Hz
Product Code. 12176640 Shop All Eppendorf Products
Product Code. 12176640 Supplier Eppendorf™ Supplier No. 6332000010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Eppendorf Mastercycler nexus enables the user to choose the consumable that best fits their needs as it can accommodate 96-well PCR plates, 0.2 mL PCR tubes, 0.2 mL PCR tube strips, and 0.5 mL PCR tubes. As well as being highly flexible, the instrument is extremely quiet and energy efficient.
Mastercycler™ nexus eco, 230 V/50 – 60 Hz- Small footprint- Universal block for ultimate flexibility- E-mail notification- flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
TRUSTED_SUSTAINABILITY
Specifications
Thermal Accuracy ±0.2°C
Thermal Uniformity ≤±0.3°C
Dimensions 41.2 x 25 x 32.1 cm
Capacity 96 x 0.2 mL PCR Tubes, 1 PCR Plate (96-wells) or up to 71 x 0.5 mL PCR Tubes
Voltage 230 V
Electrical Requirements 230 V, 50/60 Hz
Product Type PCR
Thermal Range 4°C to 99°C
For Use With (Application) Standard PCR, Fast PCR, In Situ PCR
Frequency 50 to 60 Hz
User Interface Intuitive
Wattage 540 W
Max. Ramp Rate 3°C/s (Measured at the Block)
No. of Programs >700
Heated Lid Yes
Weight 10.9 kg
Block Configurations 0.2 mL Tubes, 0.5 mL Tubes, 96-well Plate