Learn More
Atago™ MASTER-S/MillM Model Salinity Refractometer
Brand: Atago™ 2493
Additional Details : Weight : 0.10000kg
Specifications
Salinity Refractometer | |
MASTER-S/MillM | |
Measures the salinity and specific gravity of seawater, and utilizes a new material that is durable against salty samples |
0 to 100 Salinity, 1.000 to 1.070 Specific Gravity | |
Salinity, Specific gravity | |
110 g |