Learn More
Masterflex™ Mass Flowmeter
Product Code. 15128016 Shop All Masterflex Products
Change view
Click to view available options
Connections:
1/4 in. Compression Fittings
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections Unit Size
15128016 1/4 in. Compression Fittings Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15128016 Supplier Masterflex™ Supplier No. 3270732
  • ONLINE EXCLUSIVE PRICE
    1225.00 EUR
    934.14 EUR / Pack of 1
    Save 290.86 EUR
    23% Off
Estimated Shipment: 22-02-2027
Economical, highly accurate, and rapid flowmeter with a miniature coil design. Cole-Parmer™ Mass Flowmeter provides precise and repeatable measurements.
  • Thermal gas flow sensing technique
  • Calibrated to air/nitrogen
  • Cable hub connector for a quick connection to a power supply and 0 to 5 VDC output cable; order power supplies/cables under separately
  • Low pressure drop across the sensor
  • Output signal can connect to a remote display, recorder, or any instrument that accepts a 0 to 5 VDC signal
  • Compact size makes it easy to carry
  • Wetted materials are anodized aluminum, 316 stainless steel, Viton™ O-rings, and acetal fittings.
Specifications
Type Mass Flowmeter
Accuracy ±1.5% (Full Scale)
Gas Type Air, Nitrogen
Pressure Rating 150 psig
Connections 1/4 in. Compression Fittings
Display 3.5-digit LCD
Flow Rate Range 2 LPM
Repeatability ±0.5% (Full Scale)
Temperature (Metric) 10°C to 50°C