Learn More
Emma Safety Footwear Martino (Frontier 162) Safety Shoes
Product Code. 15805781
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15805781
Brand: Emma Safety Footwear MM16209042
The executive safety shoe is made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2 tone full grain leather combined with nubuck adds to the stylish look.
  • S3-Safety shoe with sole plate for wet work
  • Thermoplastic Polyurethane (TPU) Outsole
  • Full grain leather construction
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Brown
Design Martino (Frontier 162)
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 42
Toe Material Steel