Learn More
MAPK13-IN-1, MedChemExpress
Description
MPAK13-IN-1 is a MAPK13 (p38δ) inhibitor, with an IC50 of 620 nM.
Specifications
Specifications
| Chemical Name or Material | MAPK13-IN-1 |
| CAS | 229002-10-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H23N5O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 250 mg/mL (684.13 mM) |
| SMILES | O=C(NC1=CC=C(OC2=CC=NC=C2)C=C1)NC3=CC(C(C)(C)C)=NN3C |
| Molecular Weight (g/mol) | 365.43 |
| Formula Weight | 365.43 |
| Show More |