missing translation for 'onlineSavingsMsg'
Learn More

Manganese, Sigma-Aldrich™

Product Code. 15857900
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15857900 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15857900 Supplier Sigma Aldrich Supplier No. 1122370250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Powder 99+

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-96-5
Melting Point 1246°C
Boiling Point 2061°C
Density 7.4 g/cm3 at 19°C
Quantity 250 g
Form Powder
MDL Number MFCD00011111
Synonym Manganese, Manganese
Molecular Weight (g/mol) 54.94
Chemical Name or Material Manganese
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.