missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Chloride Tetrahydrate, Supelco™

Product Code. 15837410
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15837410 1 kg 1kg
15827410 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15837410 Supplier Supelco Supplier No. 1059271000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9

Specifications

Boiling Point 1190°C/1013 hPa (anhydrous substance)
Melting Point 650°C (anhydrous substance)
pH 3.5 to 6 (25°C, 50 g/L in H2O)
Quantity 1 kg
MDL Number MFCD00149792
Synonym Manganese(II) chloride tetrahydrate, Manganese dichloride, Manganese(II) chloride tetrahydrate
Molecular Weight (g/mol) 197.91
Percent Purity 99.0 to 101.0% (complexometric)
Grade Research
Density 2.01 g/cm3 at 20°C
Chemical Name or Material Manganese(II) Chloride Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.