Learn More
Manganese Dioxide, MP Biomedicals
Product Code. 11423126 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423126 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11423126 Supplier MP Biomedicals Supplier No. 0522201425
  • ONLINE EXCLUSIVE PRICE
    57.35 EUR
    54.60 EUR / 25g
    Save 2.75 EUR
    4% Off
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity powder for use in chemical research and lithium insertion studies.
Specifications
CAS 1313-13-9
Physical Form Powder
Quantity 25 g
Molecular Formula MnO2
MDL Number MFCD00003463
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILES O=[Mn]=O
IUPAC Name dioxomanganese
Molecular Weight (g/mol) 86.94
PubChem CID 14801
Formula Weight 86.94
Grade Research
Chemical Name or Material Manganese Dioxide