Description
Quick and easy assembly and disassembly without the need for tools. Barbed ridges provide secure grip on the inside of flexible tubing. Slip luer fittings allow for a simple yet secure connection
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diameter (English) Inner | 3/32 in. |
| Inlet | Male Luer Slip, 200 Series Barb |
| Material (Seal) | Polypropylene |