Learn More
Thermo Scientific™ Male Luer-LOK Priming Syringes
Product Code. 13436886 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Product Type:
GC Autosampler Priming Syringe, Luer-Lok
GC Autosampler Syringe
Gas Tight GC Autosampler Syringe, Luer-Lok
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Product Type Unit Size
13436886 10 mL GC Autosampler Syringe Each
10524655 1 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10409254 2.5 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10137854 5 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10585995 10 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10409444 25 mL Gas Tight GC Autosampler Syringe, Luer-Lok Each
10351305 50 mL GC Autosampler Priming Syringe, Luer-Lok Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 13436886 Supplier Thermo Scientific™ Supplier No. 365N2721
 Restricted Item
Estimated Shipment: 25-02-2027
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
  • TRUSTED_SUSTAINABILITY
    Specifications
    Description 10 mL FN GT LC, 19 G, 57 mm
    Needle Gauge 19 G
    Length (Metric) 57 mm
    Product Type GC Autosampler Syringe
    Quantity Ea.
    Volume (Metric) 10 mL
    Length (Metric) Needle 57 mm
    Needle Point Style LC
    For Use With (Equipment) TriPlus RSH Autosampler
    PTFE Tipped Plunger Yes