missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Mal-amido-PEG2-C2-amido-Ph-C2-CO-AZD

Product Code. p-200104441
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473236

Brand: MedChemExpress HY113697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mal-amido-PEG2-C2-amido-Ph-C2-CO-AZD is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.

Specifications

Molecular Weight (g/mol) 528.55 g/mol
CAS 1037589-69-7
Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Quantity 25 mg
Solubility DMSO : 100 mg/mL (189.20 mM; Need ultrasonic)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.