Learn More
Invitrogen™ MagniSort™ Magnet
Product Code. 15552567
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15552567 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This magnet generates a strong magnetic field. Keep away from pacemakers, credit cards, magnetic I.D. cards, watches, computer monitors and hard disks to prevent damage to these devices.
Product Code. 15552567 Supplier Invitrogen™ Supplier No. MAG490210
  • ONLINE EXCLUSIVE PRICE
    1056.00 EUR
    791.65 EUR / 5mL
    Save 264.35 EUR
    25% Off
 Restricted Item
Estimated Shipment: 26-11-2026
This magnet generates a strong magnetic field. Keep away from pacemakers, credit cards, magnetic I.D. cards, watches, computer monitors and hard disks to prevent damage to these devices.
Designed to be used with 12 x 75mm round bottom polystyrene tubes (5mL), and can be used to magnetically separate up to 2 x 108 cells. For detailed instructions, follow the protocols provided with each separation kit.
This magnet is composed of strong rare earth magnets. Rare earth magnets are sensitive to heat above 175°F (80°C). Keep away from extreme heat source.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Magnet
For Use With (Equipment) 12 x 75 mm round bottom polystyrene tubes
Quantity 5 mL