missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Carbonate, Pentahydrated Basic, Extra Pure, Heavy, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical

Product Code. 10626842 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10626842 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10626842 Supplier Fisher Chemical Supplier No. M/0450/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 12125-28-9
Color White
Physical Form Solid
Quantity 500 g
Molecular Formula CMgO3·1/4H2O2Mg·5/4H2O
MDL Number 149784
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.