missing translation for 'onlineSavingsMsg'
Learn More

Magnesium bis(trifluoromethylsulfonyl)imide

Product Code. p-7080173
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412747 250 mg 250mg
15492737 1 g 1g
15402747 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412747 Supplier Thermo Scientific Alfa Aesar Supplier No. 046667.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 133395-16-1 | C4F12MgN2O8S4 | 584.576 g/mol

Magnesium bis(trifluoromethylsulfonyl)imide is a strong Lewis acid and used as a catalyst in organic synthesis. It is an electrolyte, which shows high anodic stability used in rechargeable magnesium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133395-16-1
Molecular Formula C4F12MgN2O8S4
Molecular Weight (g/mol) 584.576
MDL Number MFCD01321278
InChI Key DMFBPGIDUUNBRU-UHFFFAOYSA-N
Synonym bis bis trifluoromethylsulfonyl amino magnesium, magnesium bis trifluoromethanesulfonimide, magnesium ii bis trifluoromethanesulfonyl imide, magnesium ii triflimide, magnesium bistrifluoromethanesulfonimide, magnesium bis trifluoromethylsulfonyl imide, magnesium 2+ bis bistriflylimide anion, bis trifluoromethanesulfonyl imide magnesium ii salt
PubChem CID 11851070
IUPAC Name magnesium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Mg+2]

Specifications

Melting Point >200°C
Color White to Yellow
Linear Formula [(CF3SO2)2N]2Mg
Quantity 250 mg
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 584.58
Physical Form Solid
Chemical Name or Material Magnesium bis (trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.