Learn More
Applied Biosystems™ MagMAX™ Cell and Tissue DNA Extraction Buffer
Product Code. 16584810
Change view
Click to view available options
No. of Reactions:
100 Preps
1000 Preps
Unit Size:
1000 preps
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
16584810 100 Preps Each
17165772 1000 Preps 1000 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 16584810 Supplier Applied Biosystems™ Supplier No. A45469
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.
MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 60 mL Cell and Tissue DNA Extraction Buffer
Sample Type Cells, Tissue
For Use With (Application) Next-Generation Sequencing, PCR, Real-Time PCR
High-throughput Compatibility Automated Protocols, High-throughput Compatible, Manual Protocols
No. of Reactions 100 Preps
Product Line MagMAX
Product Type Cell and Tissue DNA Extraction Buffer
Quantity 100 Preps
Shipping Condition Room Temperature
Target Genomic DNA