Specifications
| Product Type | Universal Platform |
| Material | Steel |
| Dimensions (L x W) | 45 x 45 cm (17.8 x 17.8 in.) |
| Description | Magnetic Attachment |
| For Use With (Application) | Holding racks and clamps |
| For Use With (Equipment) | Shaking Incubator 16333226 |