Learn More
MP Biomedicals™ Magbeads FastDNA Kit for FFPE
Product Code. 17772448 Shop All MP Biomedicals Products
Change view
Click to view available options
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17772448 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17772448 Supplier MP Biomedicals™ Supplier No. 116576096
  • ONLINE EXCLUSIVE PRICE
    496.00 EUR
    472.50 EUR / 1 sets
    Save 23.50 EUR
    4% Off
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
MagBeads FastDNA Kit for FFPE is intended for rapid extraction of DNA from FFPE sample. This kit uses two combination methods. High-salt Bind is conducive to remove pigments or polysaccharides from complex FFPE samples, so as to improve the purity of nucleic acid and avoid blocking aligent 2100.
MagBeads FastDNA Kit for FFPE is based on the purification method of high binding magnetic particles:
  • The sample is lysed and digested
  • DNA is released into the lysate
  • After addition of magnetic particles and binding solution, DNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
  • The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts
  • Finally the DNA was eluted with Elution Buffer
TRUSTED_SUSTAINABILITY
Specifications
Format Kit
Isolation Technology Magnetic Bead
Scale Mini
Sample Type FFPE
Sequencing Type DNA and RNA Sequencing
No. of Reactions 96 Preps
compliance-icons compliance-icons
Product Identifier
  • Magbeads FastDNA Kit for FFPE
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Proteinase K