Learn More
Hellma Macro Cuvette, 402-OG
Product Code. 10065991
Change view
Click to view available options
Capacity (Metric):
30,000 μL
6000 μL
Path Length:
10 mm
50 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Path Length Unit Size
10065991 6000 μL 10 mm Each
10341692 30,000 μL 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10065991 Supplier Hellma Supplier No. 4021010
  • ONLINE EXCLUSIVE PRICE
    94.45 EUR
    92.24 EUR / Pack of 1
    Save 2.21 EUR
    2% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
Optical glass absorption assay cuvette with a PTFE lid.
  • Optical glass construction
  • VIS range
  • PTFE lid included
Specifications
Capacity (Metric) 6000 μL
Material Optical Glass
Closure Material PTFE
Width (Metric) Interior 18.5 mm
Thickness (Metric) Base thickness: 2.5 mm
Includes PTFE Lid
Quantity 1/Pk
Path Length 10 mm
Wavelength Range 360 to 2500 nm
Height (Metric) 40 mm
Width (Metric) 23.6 mm