Learn More
Hellma Macro cell 110-QS, quartz glass SUPRASIL
Product Code. 10748141
Change view
Click to view available options
Path Length:
10 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Path Length Unit Size
10748141 10 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10748141 Supplier Hellma Supplier No. 1101040
  • ONLINE EXCLUSIVE PRICE
    189.00 EUR
    181.41 EUR / Pack of 1
    Save 7.59 EUR
    4% Off
Estimated Shipment: 19-10-2026
Volume: 3500 μL
Specifications
Capacity (Metric) 3500 μL
Closure Material PTFE
Closure Color White
Path Length 10 mm
Wavelength Range 200 to 2500 nm