Learn More
Emma Safety Footwear Mack M Safety Shoes
Product Code. 15998671
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15998671 37 1 pair
15908681 38 1 pair
15918681 39 1 pair
15928681 40 1 pair
15938681 41 1 pair
15948681 42 1 pair
15958681 43 1 pair
15968681 44 1 pair
15978681 45 1 pair
15988681 46 1 pair
15998681 47 1 pair
15908691 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15998671 Supplier Emma Safety Footwear Supplier No. MM53486337
  • ONLINE EXCLUSIVE PRICE
    169.05 EUR
    104.84 EUR / 1 pairs
    Save 64.21 EUR
    37% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
A superb shoe for galvanizing and welding metal. A closed front flap protects against stray sparks and hot metal splinters and offers enhanced protection for the metatarsal. Includes polyurethane outer toe protection and a concealed steel toe cap.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Heat and Cold insulated
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Mack M
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 37
Toe Material Steel