missing translation for 'onlineSavingsMsg'
Learn More

Macitentan, MedChemExpress

Product Code. 16496406
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496406 100 mg 100mg
16455988 10 mg 10mg
16448607 5 mg 5mg
16402137 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496406 Supplier Medchem Express Supplier No. HY14184
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Macitentan (ACT-064992) is an orally active, non-peptide dual ETA and ETB (endothelin receptor) antagonist. Macitentan has the potential for idiopathic pulmonary fibrosis (IPF) and pulmonary arterial hypertension (PAH).

Macitentan (ACT-064992) is an orally active, non-peptide dual ETA and ETB (endothelin receptor) antagonist. Macitentan has the potential for idiopathic pulmonary fibrosis (IPF) and pulmonary arterial hypertension (PAH).

Specifications

Chemical Name or Material Macitentan
CAS 441798-33-0
Color White
Purity Grade Notes Research
Molecular Formula C19H20Br2N6O4S
Quantity 100 mg
Synonym ACT-064992
Solubility Information DMSO : ≥ 50 mg/mL (84.99 mM)
SMILES BrC1=CN=C(OCCOC2=C(C3=CC=C(Br)C=C3)C(NS(NCCC)(=O)=O)=NC=N2)N=C1
Molecular Weight (g/mol) 588.27
Formula Weight 588.27
Percent Purity 98.75%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.