missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylhydrazine, 97%, Thermo Scientific™

Product Code. p-7045779
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328525 2 g 2g
11338525 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328525 Supplier Thermo Scientific Alfa Aesar Supplier No. L11958.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
m-Tolylhydrazine is used as pharmaceutical intermediate. It reacts with cyclohexanone and obtain the 7-methyl-1,2,3,4-tetrahydro-carbazole.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-89-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00047815
InChI Key GPTOGZLZMLJZCV-UHFFFAOYSA-N
Synonym m-tolylhydrazine, 3-methylphenyl hydrazine, hydrazine, 3-methylphenyl, m-tolylhydrazin, m-toluidine amine, m-tolyl hydrazine, m-tolyl-hydrazine, 1-m-tolylhydrazine, 3-methylphenylhydrazinehydrochloride, acmc-20akdk
PubChem CID 68308
IUPAC Name (3-methylphenyl)hydrazine
SMILES CC1=CC=CC(NN)=C1

Specifications

Density 1.057
Boiling Point 132°C to 134°C (16 mmHg)
Refractive Index 1.589
Quantity 2 g
UN Number UN2810
Beilstein 742264
Merck Index 14,9542
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material m-Tolylhydrazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.