Learn More
m-Toluidine, Sigma-Aldrich™
Product Code. 15859600
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859600 1 L 1 L
15849600 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15859600 Supplier Sigma-Aldrich Supplier No. 8083141000
 Restricted Item
Estimated Shipment: 11-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-44-1
Specifications
Melting Point -30°C
Density 0.99 g/cm3 at 20°C
Boiling Point 204°C/1013 hPa
Molecular Formula 3-(CH3)C6H4NH2
MDL Number MFCD00007808
Quantity 1 L
Synonym m-Toluidine, 3-Methylaniline, 3-Aminotoluene, m-Toluidine
Molecular Weight (g/mol) 107.15
Grade Research