Learn More
Gilson™ M-384 Well Adapter with Positioning by Hand
Product Code. 15803192 Shop All Gilson Products
Change view
Click to view available options
For Use With (Equipment):
PLATEMASTER™
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15803192 PLATEMASTER™ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15803192 Supplier Gilson™ Supplier No. F1077603
  • ONLINE EXCLUSIVE PRICE
    1020.00 EUR
    940.80 EUR / Pack of 1
    Save 79.20 EUR
    7% Off
In Stock
An accessory allowing for the use of 384-well microplates with positioning done by hand.
This adapter is an accessory for the PLATEMASTERTM that allows use of 384-well microplates with positioning done by hand.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pipetter Accessory
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001
Color Gray
Material Polypropylene
For Use With (Application) Liquid Handling
For Use With (Equipment) PLATEMASTER™
Quantity 1
Warranty 1 Year
Width (English) 6.26 in
Height (English) 2.99 in
Height (Metric) 76 mm
Weight (English) 2.21 lb
Weight (Metric) 1 kg
No. per Pack 1