Learn More
Invitrogen™ LyticBLAzer™-FRET B/G Homogeneous Kit
Product Code. 11514936
Change view
Click to view available options
Quantity:
150 mL
3.75 L
Unit Size:
150mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11514936 150 mL 150mL
10378572 3.75 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11514936 Supplier Invitrogen™ Supplier No. K1148
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LyticBLAzer™ -FRET B/G Substrate Kits provides a ratiometric, end-point method of analyzing beta-lactamase activity in cell lysates.
The LyticBLAzer™ -FRET B/G Reagent combines the components for lysis and detection in a single reagent addition, resulting in a very simple assay. Use the LyticBLAzer™ on top-read or bottom-read instrumentation. LyticBLAzer™ Substrate Kits are available in homogeneous as well as non-homogeneous formats (Figure 1), each optimized for a different assay condition.
LyticBLAzer™ FRET B/G Substrate Kit Advantages:
• Ratiometric blue:green reporter readout reduces false hits by minimizing experimental noise
• Integrated lysis and detection components enable use with top-read or bottom read fluorescence instrumentation
• Endpoint analysis and stable fluorescence yield longer read times
LyticBLAzer™ -BODIPY™ FL Substrate Kit:
LyticBLAzer™ -BODIPY™ FL Substrate Kits provides an end-point, non-ratiometric method of analyzing beta-lactamase activity in cell lysates. The LyticBLAzer™ -BODIPY™ FL Reagent combines the components for lysis and detection in a single reagent addition, resulting in a very simple assay. Use the LyticBLAzer™ -BODIPY™ FL Assays on top-read or bottom-read instrumentation. LyticBLAzer™ -BODIPY™ FL Substrate Kits are available in homogeneous as well as non-homogeneous formats (Figure 1), each optimized for a different assay condition.
LyticBLAzer™ -BODIPY™ FL Substrate Kit Advantages:
• BODIPY™ fluorophore excitation avoids fluorescence interference caused by sample contaminants such as dust, lint, etc.
• Standard excitation and emission wavelengths are universally adaptable to all fluorescence instrumentation.
• Integrated lysis and detection components enable use with top-read or bottom read fluorescence instrumentation.
• Endpoint analysis and stable fluorescence yield longer read times
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fluorometer, Microplate Reader
Product Line LyticBLAzer
Content And Storage LyticBLAzer™ homogenous kits: LyticBLAzer™-FRET B/G Homogenous Loading Kits include LyticBLAzer™_h-FRET B/G Substrate, DMSO (for Solution A), and Homogenous Lysis Buffer. LyticBLAzer™_h-FRET B/G Substrate (provided lyophilized) should be stored at -20°C, and protected from light. LyticBLAzer™_h-FRET B/G Substrate should also be desiccated. DMSO (for Solution A) and Homogenous Lysis Buffer should be stored at 22°C to 25°C, protected from direct light.
The 150 mL kit is sufficient for 3,750 x 40 μL standard protocol well assays; the 3.75 L kit is sufficient for 93,750 x 40 μL standard protocol well assays. Bulk quantities of loading kits are available upon request.
LyticBLAzer™ standard kits: LyticBLAzer™-FRET B/G Loading Kits include LyticBLAzer™-FRET B/G substrate, DMSO (for Solution A), and Lysis Buffer. LyticBLAzer™-FRET B/G substrate is provided lyophilized and should be stored at -20C, desiccated and protected from light. DMSO (for Solution A) and Lysis Buffer should be stored at 22°C to 25°C, protected from direct light.
The 150 mL kit is sufficient for 3,750 x 40 μL standard protocol well assays; the 3.75 L kit is sufficient for 93,750 x 40 μL standard protocol well assays. Bulk quantities of loading kits are available upon request.
LyticBLAzer™_h-BODIPY™ FL Substrate, DMSO (for Solution A), and Homogenous Lysis Buffer. LyticBLAzer™_h-BODIPY™ FL Substrate (provided lyophilized) should be stored at -20C, and protected from light. LyticBLAzer™_h-BODIPY™ FL Substrate should also be
desiccated. DMSO (for Solution A) and Homogenous Lysis Buffer should be stored at 22°C to 25°C, protected from direct light.
LyticBLAzer™-BODIPY™ FL Loading Kits include LyticBLAzer™-BODIPY™ FL substrate, DMSO (for Solution A), and Lysis Buffer. LyticBLAzer™-BODIPY™ FL substrate is provided lyophilized and should be stored at -20°C, desiccated and protected from light. DMSO (for Solution A) and Lysis Buffer should be stored (at 22°C to 25°C, protected from direct light.)
Detection Method Fluorescence
Product Type Kits for Analysis of Beta-Lactamase Activity
Quantity 150 mL
Format Kit
Substrate Beta-Lactamase Substrate