missing translation for 'onlineSavingsMsg'
Learn More

LUT014, MedChemExpress

Product Code. 16475396
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475396 100 mg 100mg
16470847 50 mg 50mg
16444548 10 mg 10mg
16407267 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475396 Supplier Medchem Express Supplier No. HY111940

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LUT014 is a B-Raf inhibitor with an IC50 of 11.7 nM, and developed to reduce dose-limiting acneiform lesions associated EGFR Inhibitors treatment. Extracted from patent WO 2019026065A2 .

LUT014 is a B-Raf inhibitor with an IC50 of 11.7 nM, and developed to reduce dose-limiting acneiform lesions associated EGFR Inhibitors treatment. Extracted from patent WO 2019026065A2 .

Specifications

Chemical Name or Material LUT014
CAS 2274819-46-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H19F3N8O
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (94.61 mM; Need ultrasonic)
SMILES FC(F)(F)OC1=CC(NC2=NC=CC3=C2C=CC(C)=C3NC4=NC=CC=C4C5=C6N=CNC6=NC=N5)=CC=C1
Molecular Weight (g/mol) 528.49
Formula Weight 528.49
Percent Purity 97.23%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.