missing translation for 'onlineSavingsMsg'
Learn More

Lurasidone, MedChemExpress

Product Code. 16488328
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488328 10 mg 10mg
16434217 50 mg 50mg
16408076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488328 Supplier Medchem Express Supplier No. HYB0032A
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lurasidone (SM-13496) is an antagonist of both dopamine D2 and 5-HT7 with IC50s of 1.68 and 0.495 nM, respectively. Lurasidone (SM-13496) is also a partial agonist of 5-HT1A receptor with an IC50 of 6.75 nM.

Lurasidone (SM-13496) is an antagonist of both dopamine D2 and 5-HT7 with IC50s of 1.68 and 0.495 nM, respectively. Lurasidone (SM-13496) is also a partial agonist of 5-HT1A receptor with an IC50 of 6.75 nM.

Specifications

Chemical Name or Material Lurasidone
CAS 367514-87-2
Color White
Purity Grade Notes Research
Molecular Formula C28H36N4O2S
Quantity 10 mg
Synonym SM-13496
Solubility Information DMSO : 20.83 mg/mL (42.28 mM; Need ultrasonic) ∣Ethanol : 3.33 mg/mL (6.76 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C([C@H]1[C@H]2C[C@H](CC2)[C@@H]31)N(C[C@H](CCCC4)[C@@H]4CN5CCN(CC5)C6=NSC7=CC=CC=C67)C3=O
Molecular Weight (g/mol) 492.68
Formula Weight 492.68
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.