Learn More
Thermo Scientific™ Luer-Tip, Gas-Tight Manual Syringes for GC Systems
Product Code. 15214603 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
1000 μL
10 mL
For Use With (Equipment):
Gas Chromatography Systems
TriPlus RSH Autosampler
TriPlus RSH, TriPlus 100LS Autosamplers
Unit Size:
Each
Needle Gauge:
23s G
26 G
26s G
Length (Metric):
57 mm
80 mm
85 mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) For Use With (Equipment) Unit Size Needle Gauge Length (Metric)
15214603 10 mL Gas Chromatography Systems Each - -
10371195 5 μL - Each 26 G 80 mm
12886026 5 μL TriPlus RSH, TriPlus 100LS Autosamplers Each 23s G 57 mm
15274593 1000 μL Gas Chromatography Systems Each - -
13416856 10 μL TriPlus RSH Autosampler Each 23s G 85 mm
13446856 10 μL TriPlus RSH Autosampler Each 26s G 85 mm
13426856 10 μL TriPlus RSH Autosampler Each 23s G 85 mm
12806036 10 μL TriPlus RSH Autosampler Each 23s G 57 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15214603 Supplier Thermo Scientific™ Supplier No. 365N7820
Estimated Shipment: 11-11-2026
Get a gas-tight syringe for manual injections. This syringe features a Luer-Tip connection and is available in a variety of sizes.
Get a gas-tight syringe for manual injections in your GC system. This syringe features a Luer-Tip connection and is available in a variety of sizes. Made of quality materials, they are designed for easy, accurate, precise and reproducible injections.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gas Tight GC Autosampler Syringe, Luer Tip
Volume (Metric) 10 mL
For Use With (Equipment) Gas Chromatography Systems