missing translation for 'onlineSavingsMsg'
Learn More

Lucifer Yellow CH Dipotassium Salt (Technical Grade), TRC

Product Code. 30179453
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179453 5 mg 5mg
30179158 25 mg 25mg
30178917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179453 Supplier Toronto Research Chemicals Supplier No. TRCL4737235MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71206-95-6
Molecular Formula C13H9K2N5O9S2
Molecular Weight (g/mol) 521.57
Synonym 6-Amino-2-[(hydrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-benz[de]isoquinoline-5,8-disulfonic Acid Dipotassium Salt
SMILES O=[S](OC1=CC(C)=NN1C2=CC=CC=C2)(O)=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H11N5O9S2.2K/c14-10-5-1-4(28(22,23)24)2-6-9(5)7(3-8(10)29(25,26)27)12(20)18(11(6)19)17-13(21)16-15;;/h1-3H,14-15H2,(H2,16,17,21)(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2
Chemical Name or Material Lucifer Yellow CH Dipotassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.