CAS RN 71206-95-6
CAS RN 71206-95-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lucifer Yellow CH Dipotassium Salt (Technical Grade), TRC
CAS: 71206-95-6 Molecular Formula: C13H9K2N5O9S2 Molecular Weight (g/mol): 521.57 Synonym: 6-Amino-2-[(hydrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-benz[de]isoquinoline-5,8-disulfonic Acid Dipotassium Salt SMILES: O=[S](OC1=CC(C)=NN1C2=CC=CC=C2)(O)=O